Learn More
Enzo Life SciencesSupplier Diversity Partner GLP-1 (7-37, 7-36, amide,free NT) monoclonal antibody 10 (biotin) (50 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner BPDABS03310B005
Clone: 10. Host: Mouse. Isotype: IgG2aκ. Immunogen: Synthetic peptide corresponding to aa 7-17 of GLP-1. UniProt: P01275. Crossreactivity: Shows <0.2% cross-reactivity with GLP-1(9-36)amide, glucagon and human GIP. Cross-reacts 0.25% with GLP-1(1-37), approximately 6% with human GLP-2 and approximately 50% with exendin-4. Applications: ELISA. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.14mM sodium chloride and 15mM sodium azide.
Catalog No. 50-658-899