Learn More
GLP1 Mouse anti-Human, Mammalia, Janelia Fluor 549, Clone: 5B10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP105189JF549
Catalog No. NBP15189549
Add to cart
Description
Description
GLP1 Monoclonal antibody specifically detects GLP1 in Human, Mammalia samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
Specifications
GLP1
5B10
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
GCG
Synthetic GLP-1(7-36)amide coupled to carrier and adsorbed onto aluminum hydroxide gel.
Protein A or G purified
Cancer, GPCR, Signal Transduction
Primary
2641
Human, Mammalia
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Reacts with all forms of GLP-1, including precursor and GLP-1(9-37)GLP-1(9-36 amide) metabolite.