Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP1 Mouse anti-Human, Mouse, Mammalia, Janelia Fluor 646, Clone: 8G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223506JF646

Catalog No. NBP23506646


Add to cart

Description

Description

GLP1 Monoclonal antibody specifically detects GLP1 in Human, Mouse, Mammalia samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

GLP1
8G9
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
GCG
Synthetic GLP-1(7-36)amide coupled to carrier and adsorbed onto aluminum hydroxide gel
Protein A or G purified
Cancer, GPCR, Signal Transduction
Primary
2641
Human, Mammalia, Mouse
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Clone 8G9 is specific for the amidated C-terminus of the peptide and does not react with GLP-1(7-37)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit