Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP1 Mouse anti-Human, Mammalia, Clone: 5B10, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | GLP1 |
---|---|
Clone | 5B10 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:1000, ELISA 1:20000, Immunohistochemistry 1:3200, Immunocytochemistry/Immunofluorescence 1:4000, Immunohistochemistry-Paraffin 1:3200 |
Applications | Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018351
|
Novus Biologicals
NBP1051890.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
GLP1 Monoclonal antibody specifically detects GLP1 in Human, Mammalia samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
GLP1 | |
1.0 mg/mL | |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Cancer, GPCR, Signal Transduction | |
P01275 | |
2641 | |
Synthetic GLP-1(7-36)amide coupled to carrier and adsorbed onto aluminum hydroxide gel. | |
Primary | |
Store at 4C. Do not freeze. |
5B10 | |
Western Blot 1:1000, ELISA 1:20000, Immunohistochemistry 1:3200, Immunocytochemistry/Immunofluorescence 1:4000, Immunohistochemistry-Paraffin 1:3200 | |
Monoclonal | |
Purified | |
RUO | |
Mammalia, Human | |
glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
GCG | |
IgG1 κ | |
Protein A or G purified | |
Reacts with all forms of GLP-1, including precursor and GLP-1(9-37)GLP-1(9-36 amide) metabolite. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title