Learn More
GLRA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 179511AP
Catalog No. 50-173-1895
Add to cart
Description
Description
GLRA1 is a subunit of a pentameric inhibitory glycine receptor, which mediates postsynaptic inhibition in the central nervous system. Defects in GLRA1 are a cause of startle disease (STHE), also known as hereditary hyperekplexia or congenital stiff-person syndrome. Multiple transcript variants encoding different isoforms have been found for GLRA1. Glycine receptors (GlyR) that include GLRA1 are members of the cys-loop family of ligand-gated ion channels, responsible for mediating the inhibitory effects of glycine. GLRA1 are widely distributed throughout the CNS, particularly within the hippocampus, spinal cord and brain stem. Diseases associated with GLRA1 include Hyperekplexia, Hereditary 1 and Hyperekplexia.
Specifications
Specifications
GLRA1
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07727, P23415, Q64018
GLRA1, glycine receptor, alpha 1, STHE
Rabbit
IgG
150 μL
-20°C
Primary
14654, 25674, 2741
Flow Cytometry, Western Blot
Unconjugated
GLRA1
Liquid
GLRA1
GLRA1 Fusion Protein Ag12431
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat