Learn More
Invitrogen™ GLRX5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114092
Description
Immunogen sequence: NAVVQILRLH GVRDYAAYNV LDDPELRQGI KDYSNWPTIP QVYLNGEFVG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
GLRX5 is monothiol glutaredoxin involved in the biogenesis of iron-sulfur clusters. GLRX5 required for normal iron homeostasis. GLRX5 is required for normal regulation of hemoglobin synthesis by the iron-sulfur protein ACO1.
Specifications
GLRX5 | |
Polyclonal | |
Unconjugated | |
GLRX5 | |
2310004O13Rik; 2900070E19Rik; AU020725; C14orf87; FLB4739; GLRX5; glutaredoxin 5; glutaredoxin 5 homolog; glutaredoxin 5 homolog (S. cerevisiae); glutaredoxin-related protein 5, mitochondrial; GRX5; Monothiol glutaredoxin-5; PR01238; PRO1238; PRSA; RGD1308383; SIDBA3; SPAHGC | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51218 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86SX6 | |
GLRX5 | |
Synthetic peptide directed towards the middle region of human GLRX5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.