Learn More
Research Products International CorpSupplier Diversity Partner Glass Staining Dish, Square, 20 x 20 cm DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 248940
These glass dishes are ideal for staining, destaining and fixing applications using large format blots. Dishes are molded from clear Pyrex borosilicate glass for durability and ease of cleaning. Dishes are 5cm deep and feature carrying handles for convenient handling. Safe for microwave use.
Catalog No. NC1462481