Learn More
GLS2 Mouse anti-Human, Clone: OTI6D11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809506
Catalog No. 50-166-9544
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a mitochondrial phosphate-activated glutaminase that catalyzes the hydrolysis of glutamine to stoichiometric amounts of glutamate and ammonia. This protein is functionally similar to the kidney glutaminase but is a little smaller in size. Originally thought to be liver-specific, this protein has been found in other tissues as well. At least one transcribed pseudogene has been found for this gene.
Specifications
Specifications
GLS2
Monoclonal
Unconjugated
GLS2
Lyophilized
GLS2
Full length human recombit protein of human GLS2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI6D11
PBS with 8% trehalose and no preservative; pH 7.3
Q9UI32
GA, GLS, LGA, hLGA
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
27165