Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALNT11 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548939

Catalog No. PA548939

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Polypeptide N-acetylgalactosaminyltransferase that catalyzes the initiation of protein O-linked glycosylation and is involved in left/right asymmetry by mediating O-glycosylation of NOTCH1. O-glycosylation of NOTCH1 promotes activation of NOTCH1, modulating the balance between motile and immotile (sensory) cilia at the left-right organiser (LRO). Polypeptide N- acetylgalactosaminyltransferases catalyze the transfer of an N- acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor. Displays the same enzyme activity toward MUC1, MUC4, and EA2 than GALNT1. Not involved in glycosylation of erythropoietin (EPO).
Specifications

Specifications

GALNT11
Polyclonal
PBS with 0.09% sodium azide
Q8NCW6, Q921L8
Polypeptide N-acetylgalactosaminyltransferase 11, Polypeptide GalNAc transferase 11, GalNAc-T11, pp-GaNTase 11, Protein-UDP acetylgalactosaminyltransferase 11, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 11, GALNT11
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
231050, 63917
Western Blot
Unconjugated
GALNT11
Liquid
GALNT11
KLH conjugated synthetic peptide between 110-138 amino acids from the N-terminal region of human GLT11.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit