Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLT8D2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112428
Description
GLT8D2 Polyclonal Antibody for Western Blot, ELISA

Specifications
GLT8D2 | |
Polyclonal | |
Unconjugated | |
GLT8D2 | |
1110021D20Rik; GALA4A; GLT8D2; glycosyltransferase 8 domain containing 2; glycosyltransferase 8 domain-containing protein 2; gycosyltransferase; RGD1560432; UNQ1901/PRO4347 | |
Rabbit | |
Protein G | |
RUO | |
74782, 83468 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
4.5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q640P4, Q9H1C3 | |
GLT8D2 | |
Recombinant Human Glycosyltransferase 8 domain-containing protein 2 protein (25-349AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction