Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLT8D4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549223
Description
Glycosyltransferase which elongates the O-linked glucose attached to EGF-like repeats in the extracellular domain of Notch proteins by catalyzing the addition of xylose.
Specifications
GLT8D4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
A0PJZ3 | |
GXYLT2 | |
KLH conjugated synthetic peptide between 404-432 amino acids from the C-terminal region of human GLT8D4. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GXYLT2 | |
Glucoside xylosyltransferase 2, 242n2, Glycosyltransferase 8 domain-containing protein 4, GXYLT2, GLT8D4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
727936 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction