Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human GLTPD1 His Protein
Highly purified. Generating reliable and reproducible results.
$398.00 - $1110.00
Specifications
Concentration | 0.25mg/mL |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | Phosphate buffered saline (pH7.4) containing 50% glycerol, 1mM DTT |
Gene ID (Entrez) | 80772 |
Molecular Weight (g/mol) | TMW: 26.8kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25842002
|
Novus Biologicals™
NBP2518420.02MG |
0.02mg |
Each for $398.00
|
|
|||||
NBP25184201
|
Novus Biologicals™
NBP2518420.1MG |
0.1mg |
Each for $1,110.00
|
|
|||||
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-214 of Human GLTPD1 The Recombinant Human GLTPD1 Protein is derived from E. coli. The Recombinant Human GLTPD1 Protein has been validated for the following applications: SDS-Page.Specifications
0.25mg/mL | |
Phosphate buffered saline (pH7.4) containing 50% glycerol, 1mM DTT | |
TMW: 26.8kDa | |
Ceramide-1-Phosphate Transfer Protein, CPTP, Glycolipid Transfer Protein Domain Containing 1, Glycolipid Transfer Protein Domain-Containing Protein 1 | |
Unconjugated | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
SDS-PAGE | |
80772 | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
GLTPD1 | |
Recombinant |
For research use only and is not approved for use in humans or in clinical diagnosis
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title