Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP1 Antibody (03), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221747AF532
Description
GLP1 Monoclonal specifically detects GLP1 in Mammal samples. It is validated for Western Blot, ELISA.Specifications
GLP1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
GCG | |
Synthetic GLP-1(9-37) adsorbed onto aluminum hydroxide gel. | |
Primary | |
binds GLP-1(1-37), GLP-1(7-37) and GLP-1(9-37). Cross-reaction with C-terminally amidated GLP-1 is 0-16% in various circumstances. | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
3 | |
Western Blot, ELISA | |
glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
Mouse | |
0.1 mL | |
2641 | |
Human | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction