Learn More
Glucagon Mouse anti-Human, Alexa Fluor™ 750, Clone: 3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221747AF750
Catalog No. NP221747A70
Add to cart
Description
Description
Glucagon Monoclonal antibody specifically detects Glucagon in Human samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
Glucagon
3
Western Blot, ELISA
glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
binds GLP-1(1-37), GLP-1(7-37) and GLP-1(9-37). Cross-reaction with C-terminally amidated GLP-1 is 0-16% in various circumstances.
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
GCG
Synthetic GLP-1(9-37) adsorbed onto aluminum hydroxide gel.
0.1 mL
Primary
2641
Human