Learn More
Glucagon Mouse anti-Human, Mammal, Alexa Fluor™ 594, Clone: 1100, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223561AF594
Catalog No. NP22361A94
Add to cart
Description
Description
Glucagon Monoclonal antibody specifically detects Glucagon in Human,Mammal samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
Glucagon
1100
Western Blot, ELISA
glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Reacts with all forms of GLP-1, including precursor and GLP-1(9-37)/GLP-1(9-36amide) metabolite
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
GCG
Synthetic GLP-1(7-36)amide coupled to carrier and adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
2641
Human, Mammalia