Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon Mouse anti-Human, Mouse, Mammal, Alexa Fluor™ 350, Clone: 8G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223506AF350

Catalog No. NP22306A30


Add to cart

Description

Description

Glucagon Monoclonal antibody specifically detects Glucagon in Human,Mouse,Mammal samples. It is validated for ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,ELISA Detection.
Specifications

Specifications

Glucagon
8G9
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Sandwich ELISA Detection
glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone 8G9 is specific for the amidated C-terminus of the peptide and does not react with GLP-1(7-37)
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
GCG
Synthetic GLP-1(7-36)amide coupled to carrier and adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
2641
Human, Mammalia, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit