Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLP1 Mouse anti-Human, Mouse, Mammalia, Alexa Fluor 532, Clone: 4, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP223558AF532
Description
GLP1 Monoclonal specifically detects GLP1 in Human, Mouse, Mammal samples. It is validated for ELISA, Immunohistochemistry.Specifications
GLP1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
GCG | |
Synthetic GLP-1(7-17) adsorbed onto aluminum hydroxide gel | |
Primary | |
This antibody binds the free N-terminus of GLP-1(7-37) and GLP-1(7-36)amide and shows <0.2% cross-reactivity with GLP-1(1-37), GLP-1(9-36)amide, human GIP and exendin-4. Cross-reacts approximately 1% with human GLP-2. | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry | |
4 | |
ELISA, Immunohistochemistry | |
glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP | |
Mouse | |
0.1 mL | |
2641 | |
Human, Mouse, Mammalia | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title