Learn More
Glucagon Mouse anti-Human, Rat, Porcine, Alexa Fluor™ 532, Clone: 9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221803AF532
Catalog No. NP221803A32
Add to cart
Description
Description
Glucagon Monoclonal antibody specifically detects Glucagon in Human,Rat,Porcine samples. It is validated for ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),ELISA.
Specifications
Specifications
Glucagon
9
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA
glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human, Porcine, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
GCG
Synthetic C-terminal fragment of glucagon coupled to carrier.
0.1 mL
Primary
2641