Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucagon Mouse, Clone: U16-850, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565859
Description
Glucagon Monoclonal antibody specifically detects Glucagon in Human, Mouse, Rat samples. It is validated for Flow cytometry (Intracellular Staining)) .Specifications
Glucagon | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
0.1 mg | |
Cell Biology | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
U16-850 | |
Unconjugated | |
GLP1; GLP2; GRPP; GCG | |
Human glucagon Recombinant Protein | |
RUO | |
Primary | |
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?
Provide Content Correction