Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucagon Polyclonal Antibody
Glucagon Polyclonal Antibody
Supplier: Thermo Scientific PA589937
Description
Glucagon Polyclonal Antibody for Western Blot, IHC (P)

Specifications
Glucagon | |
Polyclonal | |
Unconjugated | |
Gcg | |
GCG; Glicentin; Glicentin-related polypeptide; GLP1; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP1/2; GLP2; GLP-2; Glu; glucagon; Glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; glucagon-like peptide-1; GRPP; IL12p35; IL12P40; Incretin hormone; OXM; OXY; Oxyntomodulin; porcine proglucagon; PPG; preproglucagon; proglucagon | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14526, 24952, 2641 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2.16 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P01275, P06883, P55095 | |
Gcg | |
Recombinant fusion protein containing a sequence corresponding to amino acids 21-89 of human Pro-glucagon (NP_0020451). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction