Learn More
Glucokinase/GCK Antibody (OTI3E3), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270822UV
Catalog No. NBP270822UV
Add to Cart
Description
Description
Glucokinase/GCK Monoclonal specifically detects Glucokinase/GCK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Glucokinase/GCK
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
GCK
Full length recombinant protein of human Glucokinase/GCK (NP_000153) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI3E3
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
EC 2.7.1, EC 2.7.1.2, FGQTL3, GK, GLK, glucokinase (hexokinase 4), hexokinase-4, Hexokinase-D, HK4pancreatic isozyme, HKIV, HXKP, LGLK, maturity onset diabetes of the young 2, MODY2
Mouse
Affinity Purified
Lipid and Metabolism, Protein Kinase
2645.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions