Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucono delta-Lactone, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
90-80-2
C6H10O6
178.14
PHOQVHQSTUBQQK-UHFFFAOYNA-N
3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
OCC1OC(=O)C(O)C(O)C1O
90-80-2
C6H10O6
178.14
PHOQVHQSTUBQQK-UHFFFAOYNA-N
3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
OCC1OC(=O)C(O)C(O)C1O

GL107, 90-80-2, C6H10O6

Spectrum™ Glucono delta-Lactone, FCC is used as a sequestrant, a leavening agent, and an acidifier. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Quantity 50 kg
CAS Max % 1
Percent Purity 99 to 100.5%