Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gluconolactone, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
90-80-2
C6H10O6
178.14
PHOQVHQSTUBQQK-UHFFFAOYNA-N
3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
OCC1OC(=O)C(O)C(O)C1O
90-80-2
C6H10O6
178.14
PHOQVHQSTUBQQK-UHFFFAOYNA-N
3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
OCC1OC(=O)C(O)C(O)C1O

G1165, 90-80-2, C6H10O6

Spectrum™ Gluconolactone, USP is an exfoliant and used to treat acne lesions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Quantity 50 kg
CAS Max % 1
Percent Purity 99 to 101%