Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucosamine Hydrochloride Dietary Supplement, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
66-84-2
C6H14ClNO5
215.63
CBOJBBMQJBVCMW-UHFFFAOYNA-N
hydrogen 2-amino-3,4,5,6-tetrahydroxyhexanal chloride
[H+].[Cl-].NC(C=O)C(O)C(O)C(O)CO
66-84-2
C6H14ClNO5
215.63
CBOJBBMQJBVCMW-UHFFFAOYNA-N
hydrogen 2-amino-3,4,5,6-tetrahydroxyhexanal chloride
[H+].[Cl-].NC(C=O)C(O)C(O)C(O)CO

G1094, 66-84-2, C6H13NO5•HCl

Spectrum™ Glucosamine Hydrochloride, USP Dietary Supplement
Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.01
Quantity 500 g
Specific Rotation +70.0° to +73.0
CAS Max % 1
Percent Purity 98 to 102%