Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose 1-dehydrogenase Mouse anti-Human, Monkey, PE-Cyanine7, Clone: OTI2A7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271478PECY7

Catalog No. N271478PEC7


Add to cart

Description

Description

Glucose 1-dehydrogenase Monoclonal antibody specifically detects Glucose 1-dehydrogenase in Human, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

Glucose 1-dehydrogenase
OTI2A7
Flow Cytometry
Affinity Purified, Conjugated
H6PD
Full length recombinant protein of human Glucose 1-dehydrogenase (NP_004276) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
6-phosphogluconolactonase, DKFZp686A01246, EC 1.1.1.49, EC 2.7.4.3, EC 3.1.1.31, G6PD, H form, G6PDH, GDH/6PGL endoplasmic bifunctional protein, GDHglucose 1- dehydrogenase, glucose dehydrogenase, glucose dehyrogenase, glucose-6-phosphate dehydrogenase, salivary, hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase), MGC87643
Mouse
IgG1
0.1 mL
Primary
9563.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit