Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucose 1-dehydrogenase Rabbit anti-Human, DyLight 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297731UV
Description
Glucose 1-dehydrogenase Polyclonal specifically detects Glucose 1-dehydrogenase in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Glucose 1-dehydrogenase | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
6-phosphogluconolactonase, DKFZp686A01246, EC 1.1.1.49, EC 2.7.4.3, EC 3.1.1.31, G6PD, H form, G6PDH, GDH/6PGL endoplasmic bifunctional protein, GDHglucose 1- dehydrogenase, glucose dehydrogenase, glucose dehyrogenase, glucose-6-phosphate dehydrogenase, salivary, hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase), MGC87643 | |
Produced in rabbits immunized with E. coli-derived Human Glucose 1-dehydrogenase fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
9563 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title