Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose-6-phosphate dehydrogenase, from Leuconostoc mesenteroides, ammonium sulfate suspension, Affymetrix/USB™

Click to view available options
Quantity:
1kU,1kU
5kU,5kU
10kU,10kU
100kU
9001-40-5
MFCD00081656
E.C.1.1.1.49
9001-40-5
MFCD00081656
E.C.1.1.1.49
Quantity 100kU
Physical Form Suspension
Chemical Name or Material Glucose-6-phosphate dehydrogenase

EINECSNumber : 232-602-6