Learn More
Glucose 6 Phosphate Dehydrogenase Mouse anti-Human, Alexa Fluor™ 350, Clone: 2H7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222125AF350
Catalog No. NP22212A30
Add to cart
Description
Description
Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Glucose 6 Phosphate Dehydrogenase
2H7
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
G6PD
Purified recombinant fragment of human Glucose 6 Phosphate Dehydrogenase expressed in E. coli. [Uniprot: P11413]
0.1 mL
Primary
2539