Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose 6 phosphate isomerase Mouse anti-Human, Monkey, Alexa Fluor™ 647, Clone: OTI2D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270844AF647

Catalog No. N270844A647


Add to cart

Description

Description

Glucose 6 phosphate isomerase Monoclonal antibody specifically detects Glucose 6 phosphate isomerase in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Glucose 6 phosphate isomerase
OTI2D2
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
GPI
Full length recombinant protein of human Glucose 6 phosphate isomerase (NP_000166) produced in HEK293T cells.
Immunogen affinity purified
Cytokine Research, Phospho Specific
Primary
2821.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit