Learn More
Crystal Chem IncSupplier Diversity Partner Glucose Assay Kit, 96-tests DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 81696
The Glucose Assay is used to quantify levels of glucose in human serum, plasma, and urine using only a 2 µL sample volume. Highlights: Easy-to-use, results in 5 minutes. Kits use a sample size of only 2 µL. For use with serum, plasma, and urine. Designed for human samples. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7911