Learn More
BioVision IncSupplier Diversity Partner PicoProbe™ Glucose Fluorometric Assay Kit: Fluorometric Assay to Quantitate Glucose in a variety of Biological Samples Highly Sensitive, High-Throughput, Rapid & Convenient. Detection limit: less than 0.5 µM. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K688100
Glucose is the main energy source for virtually all living organisms. Glucose level is a key diagnostic parameter for many metabolic disorders. Measurement of glucose can be very important in both research and drug discovery processes. Biovision’s PicoProbe™ Glucose Assay kit is simple, rapid, ultra-sensitive and suitable for high-throughput. In this assay, D-glucose is enzymatically oxidized to form a product which reacts with a colorless probe to generate the fluorescence (Ex/Em = 535/587). The fluorescence generated is directly proportional to the amount of glucose. This assay kit can detect less than 0.5 µM glucose in various biological samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9841