Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucose-Glutamic Acid Solution, APHA, Reagents
Click to view available options
Quantity:
100 mL
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Glucose-Glutamic Acid Solution, APHA |
CAS | 7732-18-5, 56-86-0, 56-86-0 |
Color | Colorless |
Boiling Point | 100°C |
Packaging | Glass Bottle |
Quantity | 500 mL |
Specific Gravity | 1.0001 |
Grade | Reagent |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction