Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Glucose-Regulated Protein 94 (Grp94)/gp96 Ab-1, Rat Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Specifications

Antigen Glucose-Regulated Protein 94 (Grp94)/gp96 Ab-1
Clone 9G10.F8.2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RT102P
View Documents
Thermo Scientific™
RT102P
Purified Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
RT102R1
View Documents
Thermo Scientific™
RT102R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ Glucose-Regulated Protein 94 (Grp94)/gp96 Ab-1, Rat Monoclonal Antibody.

Grp's (Glucose-Regulated Proteins) are a class of proteins synthesized in cells in response to glucose starvation. Glucose-regulated protein 94 (grp94) or tumor rejection antigen (gp96) shows a high degree of sequence homology with the heat shock protein 90 (hsp90). Grp94 has been briefly studied as a prognostic factor in breast cancer.

Host Species: Rat

Clone: 9G10.F8.2

Isotype: IgG2a

Species Reactivity: Human, Monkey, Horse, Cow, Sheep, Pig, Dog, Rabbit, Rat, Mouse, Hamster, Guinea Pig, Chicken and Xenopus. Others not known.

Epitope: Not determined

Immunogen: Purified glucose-regulated protein 94 (grp94) from chicken oviducts

Molecular Weight: 94 kDa

Positive Control: SKBR-3 cells, Tonsil and Breast Carcinomas.

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native only)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Glucose-Regulated Protein 94 (Grp94)/gp96 Ab-1
Immunoprecipitation, Western Blot
Rat
Steroids Hormones and Growth Factors
Purified glucose regulated protein 94 (grp94) from chicken oviducts
9G10.F8.2
Monoclonal
RUO
Primary
IgG2a
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit