Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Glucose-Regulated Protein 94 (Grp94)/gp96 Ab-1, Rat Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Supplier:  Epredia™ RT102R1

Catalog No. RT102R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Grp's (Glucose-Regulated Proteins) are a class of proteins synthesized in cells in response to glucose starvation. Glucose-regulated protein 94 (grp94) or tumor rejection antigen (gp96) shows a high degree of sequence homology with the heat shock protein 90 (hsp90). Grp94 has been briefly studied as a prognostic factor in breast cancer.

Host Species: Rat

Clone: 9G10.F8.2

Isotype: IgG2a

Species Reactivity: Human, Monkey, Horse, Cow, Sheep, Pig, Dog, Rabbit, Rat, Mouse, Hamster, Guinea Pig, Chicken and Xenopus. Others not known.

Epitope: Not determined

Immunogen: Purified glucose-regulated protein 94 (grp94) from chicken oviducts

Molecular Weight: 94 kDa

Positive Control: SKBR-3 cells, Tonsil and Breast Carcinomas.

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native only)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Glucose-Regulated Protein 94 (Grp94)/gp96 Ab-1
Monoclonal
Unconjugated
Rat
1 mL
Steroids Hormones and Growth Factors
Bovine, Canine, Chicken, Equine, Guinea Pig, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat, Sheep
IgG2a
Immunoprecipitation, Western Blot
9G10.F8.2
Ready-to-use for IHC (UVLP)
Purified glucose regulated protein 94 (grp94) from chicken oviducts
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit