Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose Syrup, FCC, 20%, Spectrum™

Click to view available options
Quantity:
200 L
Packaging:
Drum

C3167, 8029-43-4

Spectrum™ Glucose Syrup, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Drum
Ignition Residue 0.005
Quantity 200 L
CAS 8029-43-4
CAS Max % 1
Percent Purity 20%