Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose Transporter GLUT6 Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI7E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271682JF646

Catalog No. N271682J646


Add to cart

Description

Description

Glucose Transporter GLUT6 Monoclonal antibody specifically detects Glucose Transporter GLUT6 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Glucose Transporter GLUT6
OTI7E3
Immunohistochemistry
Affinity Purified, Conjugated
SLC2A6
Full-length protein expressed in 293T cell transfected with human Glucose Transporter GLUT6 expression vector.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Glucose transporter type 6, Glucose transporter type 9, GLUT6, GLUT-9, GLUT9GLUT-6, HSA011372, solute carrier family 2 (facilitated glucose transporter), member 6, solute carrier family 2, facilitated glucose transporter member 6
Mouse
IgG2a
0.1 mL
Primary
11182.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit