Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose Transporter GLUT8 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP303554100UL

 View more versions of this product

Catalog No. NB094776


Add to cart

Description

Description

Glucose Transporter GLUT8 Polyclonal antibody specifically detects Glucose Transporter GLUT8 in Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Glucose Transporter GLUT8
Unconjugated
Purified
Rabbit
IgG
100 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Mouse, Rat
Western Blot
PBS with 50% glycerol, pH7.3.
GLUT-8, GLUT8solute carrier family 2 (facilitated glucose transporter) member 8, GLUTX1facilitated glucose transporter member 8, solute carrier family 2 (facilitated glucose transporter), member 8
Recombinant fusion protein containing a sequence corresponding to amino acids 200-260 of human Glucose Transporter GLUT8 (NP_055395.2).
Affinity purified
RUO
Primary
29988
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit