Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose6PhosphateDehydrogenase Mouse anti-Human, Janelia Fluor 549, Clone: 2F6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130358JF549

Catalog No. NBP13358549


Add to cart

Description

Description

Glucose6PhosphateDehydrogenase Monoclonal antibody specifically detects Glucose6PhosphateDehydrogenase in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Glucose6PhosphateDehydrogenase
2F6
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
G6PD
Recombinant human G6PD (35-506aa) purified from E. coli
Protein G purified
Cancer, Cellular Markers, Golgi Apparatus Markers
Primary
2539
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase
Mouse
IgG2b Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody is specific for human G6PD. This monoclonal antibody, clone 2F6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human G6PD protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit