Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Glufosinate ammonium, 95%

Click to view available options
Quantity:
250mg
1g
77182-82-2
C5H15N2O4P
198.159
MFCD00055562
ZBMRKNMTMPPMMK-UHFFFAOYSA-N
DL-Phosphinothricin; 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt
122130249
azanium;(3-amino-3-carboxypropyl)-methylphosphinate
CP(=O)(CCC(C(=O)O)N)[O-].[NH4+]
77182-82-2
C5H15N2O4P
198.159
MFCD00055562
ZBMRKNMTMPPMMK-UHFFFAOYSA-N
DL-Phosphinothricin; 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt
122130249
azanium;(3-amino-3-carboxypropyl)-methylphosphinate
CP(=O)(CCC(C(=O)O)N)[O-].[NH4+]

Interferes with the synthesis of the amino acid glutamine

Applications
Used as broad spectrum post-emergence agrochemical for grapes, orchards, plantations, ornamentals, and non-cropland; Also used as crop desiccant.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Beilstein 8163399
Merck Index 14,7339
Solubility Information Soluble in water
Formula Weight 198.16
Physical Form Powder
Percent Purity 95%
Boiling Point 519°C
Color White
Melting Point 215°C
Quantity 1g
Chemical Name or Material Glufosinate ammonium
Show More Show Less

GHS H Statement
H360-H373-H302-H312-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P260-P261-P280-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : EK7713600

storageNote1 : 2° to 8°C