Learn More
GLUL Mouse anti-Human, Clone: OTI1F4, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA500700S
Catalog No. 50-167-1842
Add to cart
Description
Description
Glutamine synthase is part of the glutamine synthetase family. Ammonia incorporation in animals occurs through the actions of glutamate dehydrogenase and glutamine synthase. Glutamate plays the central role in mammalian nitrogen flow, serving as both a nitrogen donor and nitrogen acceptor. It also has an important role in controlling metabolic regulations of neurotransmitter glutamate. Because of the multiple functions and importance of GS in cellular metabolism, both catalytic activities and synthesis are highly regulated. The activity of GS is controlled by adenylylation. Its activity is decreased in the cerebral cortex of brains affected by Alzheimer ft.s disease, particularly in the vicinity of senile plaques. It is also decreased under conditions of glucose deprivation.
Specifications
Specifications
GLUL
Monoclonal
Unconjugated
GLUL
Liquid
GLUL
Full length human recombit protein of human GLUL produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1F4
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P15104
GLNS, GS, PIG43, PIG59
Mouse
IgG2a
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2752