Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR1 (p Ser845) Rabbit anti-Human, Mouse, Rat, Clone: JJ2009, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP267853

Catalog No. NBP267853


Add to Cart

Description

Description

GluR1 Monoclonal specifically detects GluR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

GluR1 (p Ser845)
Monoclonal
1 mg/mL
Western Blot 1:500, Flow Cytometry 1:10-1:100
AMPA 1, AMPA-selective glutamate receptor 1, GluA1, GLUH1, GluR-1, GLUR1gluR-A, GluR-A, GLURA, GluR-K1, glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1, HBGR1, MGC133252
Rabbit
Protein A purified
GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Vision
2890
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Flow Cytometry, Western Blot
JJ2009
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
GRIA1
Synthetic phospho-peptide corresponding to residues surrounding Ser845 of Human GluR1 aa 831-880/ 906. (SwissProt: P42261 Human; SwissProt: P23818 Mouse; SwissProt: P19490 Rat)
100 μL
Primary
Human, Mouse, Rat
Purified
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit