Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR2 Mouse anti-Human, Rat, Primate, Clone: 3A11, Novus Biologicals™
Mouse Monoclonal Antibody
$177.15 - $666.00
Specifications
Antigen | GluR2 |
---|---|
Clone | 3A11 |
Concentration | 1.7 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:500-1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Radioimmunoassay 1:100-1:2000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays |
Description
GluR2 Monoclonal specifically detects GluR2 in Human, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Radioimmunoassay.Specifications
GluR2 | |
1.7 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays | |
Unconjugated | |
RUO | |
Human, Rat, Primate | |
AMPA 2, glutamate receptor, ionotropic, AMPA 2 | |
GRIA2 | |
IgG2a | |
Affinity Purified | |
Glutamate Receptor 2 (GluR2) by Western Blot and RIA. When using MAB396 for immunohistochemistry or immunoprecipitation it recognizes both GluR2 and GluR4. |
3A11 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:500-1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Radioimmunoassay 1:100-1:2000 | |
Monoclonal | |
Mouse | |
Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Tumor Suppressors, Vision | |
Q14416 | |
2891 | |
Recombinant fusion protein TrpE-GluR2. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title