Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR2 Rabbit anti-Human,Mouse,Rat,Bovine, KHL, Clone: 1K4, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen GluR2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB100825MI
View Documents
MilliporeSigma
ZRB100825UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB10084XMI
View Documents
MilliporeSigma
ZRB10084X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

GluR2 Monoclonal antibody specifically detects GluR2 in Human,Mouse,Rat,Bovine samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

GluR2
KHL
Rabbit
Glutamate receptor 2;AMPA-selective glutamate receptor 2;GluA2;GluR-2;GluR-B;GluR-K2;Glutamate receptor ionotropic AMPA 2
KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal cytoplasmic domain of rat Glutamate receptor 2.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat, Bovine
Gria2, Glur2
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit