Learn More
GluR3 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111782
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)EKPFHLN YHVDHLD Homology: Mouse, human, dog amino acid residues are identical.
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. The subunit encoded by this gene belongs to a family of AMPA (alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate)-sensitive glutamate receptors, and is subject to RNA editing (AGA->GGA; R->G). Alternative splicing at this locus results in different isoforms, which may vary in their signal transduction properties.
Specifications
GluR3 (extracellular) | |
Polyclonal | |
Unconjugated | |
GRIA3 | |
2900064I19Rik; AMPA-selective glutamate receptor 3; BOS_25163; GluA3; Glur3; GluR-3; Gluralpha3; GLURC; GluR-C; GluR-K3; Glutamate receptor 3; Glutamate receptor ionotropic, AMPA 3; GRIA3; Kiaa4184; MRX94; OTTHUMP00000024262; RP11-349N19.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2892, 29628, 53623 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunomicroscopy, Immunoprecipitation, Western Blot | |
0.6 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P19492, P42263, Q9Z2W9 | |
GRIA3 | |
Peptide corresponding to amino acid residues 60-73 of rat GluR3 (Extracellular, N-terminus) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.