Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR7/GRIK3 Mouse anti-Human, Clone: 2B3D1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261782

Catalog No. NBP2-61782


Add to cart

Description

Description

GluR7/GRIK3 Monoclonal antibody specifically detects GluR7/GRIK3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

GluR7/GRIK3
2B3D1
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
Purified
GRIK3
Purified recombinant fragment of human GRIK3 (AA: extra 32-173) expressed in E. Coli.
104 kDa
0.1 ml
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PBS with 0.05% Sodium Azide
EAA5, Excitatory amino acid receptor 5, GLR7, gluR-7, GluR7, GluR7a, GLUR7dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)), Glutamate receptor 7, glutamate receptor, ionotropic kainate 3, glutamate receptor, ionotropic, kainate 3
Mouse
IgG1
Protein G purified
Neuronal Cell Markers, Neurotransmission
Primary
2899
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit