Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT-1 Rabbit anti-Human,Mouse,Bovine,Rat, KHL, Clone: 16D21, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen GLUT-1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB071401MI
View Documents
MilliporeSigma
ZRB0714014X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB071425MI
View Documents
MilliporeSigma
ZRB07140125UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

GLUT-1 Monoclonal antibody specifically detects GLUT-1 in Human,Mouse,Bovine,Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blotting.
Specifications

Specifications

GLUT-1
KHL
Rabbit
Solute carrier family 2 facilitated glucose transporter member 1;Glucose transporter type 1erythrocyte/brain;HepG2 glucose transporter
KLH-conjugated linear peptide corresponding to 12 amino acids from the C-terminal region of human Solute carrier family 2, facilitated glucose transporter member 1 (GLUT-1).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Purified
Human, Mouse, Bovine, Rat
SLC2A1, GLUT1
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit