Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut1 Rabbit anti-Human, Clone: GLUT1/3132R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30788710UG

 View more versions of this product

Catalog No. NB123247


Add to cart

Description

Description

Glut1 Monoclonal antibody specifically detects Glut1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

Glut1
GLUT1/3132R
Unconjugated
10 mM PBS with 0.05% BSA
Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1
Recombinant fragment of human Glut1 protein (around aa 203-305) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)
0.2 mg/ml
Flow Cytometry 1-2 ug/million cells, ELISA 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Rabbit
IgG
100 μg
Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction
Primary
6513
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit