Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Glut1 Recombinant Protein Antigen

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

$230.50

Specifications

Product Type Recombinant Protein Antigen
Recombinant Yes
Regulatory Status RUO
Quantity 0.1mL
Storage Requirements Store at −20°C. Avoid freeze-thaw cycles.
View More Specs

Products 2
Catalog Number Mfr. No. Protein Length Price Quantity & Availability  
Catalog Number Mfr. No. Protein Length Price Quantity & Availability  
NBP248609X View Documents Novus Biologicals™
NBP248609PEP
GRTFDEIASGFRQGGASQSDKTPEELFHP Each for $230.50
Add to cart
 
NBP254745X View Documents Novus Biologicals™
NBP254745PEP
RNEENRAKSVLKKLRGTADVTHDLQE Each for $230.50
Add to cart
 
Description

Description

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human Glut1 The Glut1 Recombinant Protein Antigen is derived from E. coli. The Glut1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications

Specifications

Recombinant Protein Antigen
RUO
Store at −20°C. Avoid freeze-thaw cycles.
DYT17, DYT18, DYT18DYT17, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, Glut1
>80% by SDS-PAGE and Coomassie blue staining
Human
Blocking/Neutralizing, Control
Yes
0.1mL
SLC2A1
Glut1 Recombinant Protein Antigen
PBS and 1M Urea, pH 7.4
E.coli
Unlabeled

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit