Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut2, Rabbit anti-Human,Mouse, Janelia Fluor 549, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP222218JF549

Catalog No. NB005088


Add to cart

Description

Description

Glut2 Polyclonal specifically detects Glut2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Glut2
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2
Rabbit
IgG
0.1 mL
Lipid and Metabolism, Stem Cell Signaling Pathway
Primary
Human, Mouse
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
SLC2A2
A synthetic peptide made to an internal portion of the human GLUT2 protein (between residues 50-150) [UniProt# P11168]
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit