Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glut4 Antibody (925932), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB86541MFV500
Description
Glut4 Monoclonal antibody specifically detects Glut4 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Glut4 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cellular Markers, Core ESC Like Genes, Diabetes Research, Hematopoietic Stem Cell Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroendocrinology, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 6517 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 925932 | |
| Flow Cytometry, CyTOF-ready | |
| GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4 | |
| Mouse myeloma cell line NS0-derived recombinant human Glut4, Met1-Asp509, Accession # P14672 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction